5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: (S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, 5-Hydroxyl-L-tryptophan, (-)-5-Hydroxy-L-Tryptophane, L-5-Hydroxytryptophan, Oxitriptan, H-Trp(5-OH)-OH

H0531-1G H0531-5G
TCIAH0531-1GEA 0 CNY
TCIAH0531-1G TCIAH0531-5G
5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)
5-Hydroxy-L-tryptophan
分子式: C₁₁H₁₂N₂O₃
分子量: 220.23 g/mol
溶点: 273
存储条件: Refrigerator
MDL编号: MFCD00064341
CAS号: 4350-09-8
EINECS: 224-411-1
UN号: 2811
ADR: 6.1 III
Merck Index: 14,04847
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24fc82d09087b5b7a340b6b5bec354c2f9f515a&vid=a91f524b9dbb1f37b1a1c1ca5a7aa75d065808dd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -31.0 to -35.0 deg (C=1, H2O)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR