油酸 ≥99.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 顺式-9-十八烯酸, 十八烯酸

O0180-25ML O0180-5ML
TCIAO0180-25MLEA 0 CNY
TCIAO0180-25ML TCIAO0180-5ML
油酸 ≥99.0% (by GC, titration analysis)
油酸
分子式: C₁₈H₃₄O₂
分子量: 282.47 g/mol
沸点: 285°C
溶点: 16.7
闪点: 187°C
存储条件: Refrigerator
MDL编号: MFCD00064242
CAS号: 112-80-1
EINECS: 204-007-1
Merck Index: 14,06828
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee13c6abe07f6308fe818fe431015b08e2fdcb25&vid=ee13c6abe07f6308fe818fe431015b08e2fdcb25&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR