N,N-二甲基乙酰胺 ≥99.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAD0641-500MLEA
0
CNY
TCIAD0641-500ML
TCIAD0641-25ML
N,N-二甲基乙酰胺 ≥99.0% (通过GC)
N,N-二甲基乙酰胺
分子式:
CH₃CON(CH₃)₂ 分子量: 87,12 g/mol 沸点: 165°C 溶点: –20 °C 闪点: 66°C |
MDL编号:
MFCD00008686 CAS号: 127-19-5 EINECS: 204-826-4 ADR: 6.1,III Merck Index: 13,03254 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...