2-甲基蒽醌 ≥99.0% (by GC)
供应商: TCI
Synonyms: 2-Methylanthraquinone, 2-Methylanthracene-9,10-Dione
M0156-25G M0156-500G
TCIAM0156-25GEA 0 CNY
TCIAM0156-25G TCIAM0156-500G
2-甲基蒽醌 ≥99.0% (by GC)
2-甲基蒽醌
分子式: C₁₅H₁₀O₂
分子量: 222.24 g/mol
MDL编号: MFCD00001235
CAS号: 84-54-8
EINECS: 201-539-6
Merck Index: 14,06021
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6827fa784745139bb2ec72192e6a3a424870fa4&vid=b6827fa784745139bb2ec72192e6a3a424870fa4&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 175.0 to 178.0 deg-C