Michler 酮 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
4,4′-双(二甲氨基)二苯甲酮
B0481-25G
B0481-500G
TCIAB0481-25GEA
0
CNY
TCIAB0481-25G
TCIAB0481-500G
Michler 酮 ≥98.0% (by GC, titration analysis)
Michler 酮
分子式:
C₁₇H₂₀N₂O 分子量: 268.36 g/mol 沸点: 428 溶点: 172 闪点: 220 存储条件: Ambient |
MDL编号:
MFCD00008312 CAS号: 90-94-8 EINECS: 202-027-5 Merck Index: 14,06177 |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 173.0 to 177.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...