3-乙酰苯甲酸 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3'-Acetophenonecarboxylic acid, 3-Acetylbenzoic Acid
A1652-1G
A1652-5G
TCIAA1652-1GEA
0
CNY
TCIAA1652-1G
TCIAA1652-5G
3-乙酰苯甲酸 ≥97.0% (by GC, titration analysis)
3-乙酰苯甲酸
分子式:
C₉H₈O₃ 分子量: 164.16 g/mol 沸点: 343 溶点: 172 |
MDL编号:
MFCD00045847 CAS号: 586-42-5 |
|
产品技术规格
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 161.0 to 167.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...