1,3-二氰基苯 ≥98.0% (通过GC)

供应商: TCI
Warning

I0158-25G I0158-250G
TCIAI0158-25GEA 0 CNY
TCIAI0158-25G TCIAI0158-250G
1,3-二氰基苯 ≥98.0% (通过GC)
1,3-二氰基苯
分子式: C₈H₄N₂
沸点: 288 °C (1013 hPa)
溶点: 158…160 °C
密度: 4,42 g/cm³ (20 °C)
闪点: ≥ 150 °C
存储条件: Ambient
MDL编号: MFCD00001795
CAS号: 626-17-5
EINECS: 210-933-7
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b048936235979ba983d356434d967522fd59faa8&vid=b048936235979ba983d356434d967522fd59faa8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR