4-氨基-2-氯苯酚 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-氯-4-羟基苯胺
A1496-25G
A1496-5G
TCIAA1496-25GEA
0
CNY
TCIAA1496-25G
TCIAA1496-5G
4-氨基-2-氯苯酚 ≥98.0% (by GC, titration analysis)
4-氨基-2-氯苯酚
分子式:
C₆H₆ClNO 分子量: 143.57 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00128891 CAS号: 3964-52-1 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...