2-Amino-5-chloro-3-methylpyridine ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-Chloro-3-methyl-2-pyridylamine, 5-Chloro-3-methylpyridin-2-amine, 2-Amino-5-chloro-3-picoline

A1931-1G A1931-5G
TCIAA1931-1GEA 0 CNY
TCIAA1931-1G TCIAA1931-5G
2-Amino-5-chloro-3-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-chloro-3-methylpyridine
分子式: C₆H₇ClN₂
分子量: 142.59 g/mol
MDL编号: MFCD00129030
CAS号: 20712-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=587b601ed167c77978f2e0e684ed59d48d54f430&vid=587b601ed167c77978f2e0e684ed59d48d54f430&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR