三氯乙酰胺 ≥98.0% (by total nitrogen basis)

供应商: TCI
Warning

Synonyms: 2,2,2-Trichloroacetamide

T0368-25G T0368-500G
TCIAT0368-25GEA 0 CNY
TCIAT0368-25G TCIAT0368-500G
三氯乙酰胺 ≥98.0% (by total nitrogen basis)
三氯乙酰胺
分子式: C₂H₂Cl₃NO
分子量: 162.4 g/mol
沸点: 238
MDL编号: MFCD00008009
CAS号: 594-65-0
EINECS: 209-849-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d58f68b88bc9e6c5c1492435717ae288df83ad73&vid=d58f68b88bc9e6c5c1492435717ae288df83ad73&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 140.0 to 144.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR