2,4-二氯-6-甲基嘧啶 ≥98.0% (by GC)

供应商: TCI
Warning

D2958-25G D2958-5G
TCIAD2958-25GEA 0 CNY
TCIAD2958-25G TCIAD2958-5G
2,4-二氯-6-甲基嘧啶 ≥98.0% (by GC)
2,4-二氯-6-甲基嘧啶
分子式: C₅H₄Cl₂N₂
分子量: 163.01 g/mol
沸点: 219°C
溶点: 47
MDL编号: MFCD00006064
CAS号: 5424-21-5
EINECS: 226-563-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=025e1b224015d739fa151ac4786eb4a0283c2e1a&vid=025e1b224015d739fa151ac4786eb4a0283c2e1a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR