2,4,6-三氯嘧啶 ≥98.0% (by GC)
供应商: TCI
Warning
T0417-25G T0417-500G
TCIAT0417-25GEA 0 CNY
TCIAT0417-25G TCIAT0417-500G
2,4,6-三氯嘧啶 ≥98.0% (by GC)
2,4,6-三氯嘧啶
分子式: C₄HCl₃N₂
沸点: 210 °C (1013 hPa)
溶点: 21 °C
密度: 1,595 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00006063
CAS号: 3764-01-0
EINECS: 223-183-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc8d18919d6c4da876b871e538b96b5509567a88&vid=bc8d18919d6c4da876b871e538b96b5509567a88&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 21.0 to 23.0 deg-C