2,4,6-三甲基苯甲基氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2,4,6-三甲基氯苄

T1763-25G T1763-5G
TCIAT1763-25GEA 0 CNY
TCIAT1763-25G TCIAT1763-5G
2,4,6-三甲基苯甲基氯 ≥98.0% (by GC, titration analysis)
2,4,6-三甲基苯甲基氯
分子式: C₁₀H₁₃Cl
分子量: 168.67 g/mol
沸点: 119°C
存储条件: Refrigerator
MDL编号: MFCD00000901
CAS号: 1585-16-6
EINECS: 216-440-3
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e63d3d03909e973d4f2b71243fb6a45c6c4947&vid=02e63d3d03909e973d4f2b71243fb6a45c6c4947&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 36.0 to 42.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR