2-溴-4-甲基苯酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-溴对甲酚

B0808-25G B0808-500G
TCIAB0808-25GEA 0 CNY
TCIAB0808-25G TCIAB0808-500G
2-溴-4-甲基苯酚 ≥98.0% (by GC)
2-溴-4-甲基苯酚
分子式: C₇H₇BrO
沸点: 214°C
溶点: 26 °C
密度: 1,5468 g/cm³ (25 °C)
MDL编号: MFCD00002151
CAS号: 6627-55-0
EINECS: 229-595-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7ea286485e37219bec8a2114a5306653d1e36dc&vid=d7ea286485e37219bec8a2114a5306653d1e36dc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR