3-溴-5-碘苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

B1807-25G B1807-5G
TCIAB1807-25GEA 0 CNY
TCIAB1807-25G TCIAB1807-5G
3-溴-5-碘苯甲酸 ≥98.0% (by titrimetric analysis)
3-溴-5-碘苯甲酸
分子式: C₇H₄BrIO₂
分子量: 326.92 g/mol
MDL编号: MFCD00191851
CAS号: 188815-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed46a0ebebe5662b3fe0166f94969d9df325caba&vid=ed46a0ebebe5662b3fe0166f94969d9df325caba&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR