1-Bromo-2-chloro-4-fluorobenzene ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Bromo-3-chloro-1-fluorobenzene, 2-Chloro-4-fluorobromobenzene, 4-Bromo-3-chlorofluorobenzene

B3349-25G B3349-5G
TCIAB3349-25GEA 0 CNY
TCIAB3349-25G TCIAB3349-5G
1-Bromo-2-chloro-4-fluorobenzene ≥98.0% (by GC)
1-Bromo-2-chloro-4-fluorobenzene
分子式: C₆H₃BrClF
分子量: 209.44 g/mol
MDL编号: MFCD00142587
CAS号: 110407-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d45af4e735666560211f0e735a8d6c1f8de6356&vid=6d45af4e735666560211f0e735a8d6c1f8de6356&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR