1,3,5-三甲基吡唑 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole

T2724-25G T2724-5G
TCIAT2724-25GEA 0 CNY
TCIAT2724-25G TCIAT2724-5G
1,3,5-三甲基吡唑 ≥98.0% (by GC)
1,3,5-三甲基吡唑
分子式: C₆H₁₀N₂
分子量: 110.16 g/mol
沸点: 170°C
密度: 0.9269
MDL编号: MFCD00015536
CAS号: 1072-91-9
UN号: 1325
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e336510b711927a61fa5acf81fdc18130d84de0b&vid=e336510b711927a61fa5acf81fdc18130d84de0b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR