1,2-二甲基吲哚 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1,2-Dimethyl-1H-indole, N,2-Dimethylindole

D1391-25G D1391-5G
TCIAD1391-25GEA 0 CNY
TCIAD1391-25G TCIAD1391-5G
1,2-二甲基吲哚 ≥98.0% (by GC)
1,2-二甲基吲哚
分子式: C₁₀H₁₁N
沸点: 143°C
MDL编号: MFCD00005802
CAS号: 875-79-6
EINECS: 212-877-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c7ff5f89df464786a41f39b4045cf47abcf410b&vid=1c7ff5f89df464786a41f39b4045cf47abcf410b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR