1-苄基咪唑 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1-Benzyl-1H-imidazole

B3387-25G B3387-5G
TCIAB3387-25GEA 0 CNY
TCIAB3387-25G TCIAB3387-5G
1-苄基咪唑 ≥98.0% (by titrimetric analysis)
1-苄基咪唑
分子式: C₁₀H₁₀N₂
分子量: 158.2 g/mol
沸点: 122°C
溶点: 72
MDL编号: MFCD00005296
CAS号: 4238-71-5
EINECS: 224-200-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c314592a0951e6153aafdbd35471691608f870f6&vid=c314592a0951e6153aafdbd35471691608f870f6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR