1-乙基咪唑 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: N-Ethyl-1H-imidazole, 1-Ethyl-1H-imidazole, N-Ethylimidazole

E0132-25G E0132-250G
TCIAE0132-25GEA 0 CNY
TCIAE0132-25G TCIAE0132-250G
1-乙基咪唑 ≥98.0% (by GC, titration analysis)
1-乙基咪唑
分子式: C₅H₈N₂
分子量: 96.13 g/mol
沸点: 226°C
存储条件: Ambient
MDL编号: MFCD00055389
CAS号: 7098-07-9
EINECS: 230-403-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d41773bfc2d44fbda797dd1870b486534327ce&vid=b2d41773bfc2d44fbda797dd1870b486534327ce&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR