1,1,2,2-Tetraphenyldisilane ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: TPDS

T1896-1G T1896-5G
TCIAT1896-1GEA 0 CNY
TCIAT1896-1G TCIAT1896-5G
1,1,2,2-Tetraphenyldisilane ≥97.0% (by GC)
1,1,2,2-Tetraphenyldisilane
分子式: C₂₄H₂₂Si₂
MDL编号: MFCD00093170
CAS号: 16343-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd8773ca824b1077cf73f23dbf115caa833fde60&vid=fd8773ca824b1077cf73f23dbf115caa833fde60&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 76.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR