二苯胍 ≥98.0% (by titrimetric analysis)
供应商: TCI
Warning
Synonyms: N,N'-Diphenylguanidine, sym-Diphenylguanidine
D0892-25G D0892-500G
TCIAD0892-25GEA 0 CNY
TCIAD0892-25G TCIAD0892-500G
二苯胍 ≥98.0% (by titrimetric analysis)
二苯胍
分子式: C₁₃H₁₃N₃
沸点: 321 °C (1013 hPa)
溶点: 146…147 °C
密度: 1,19 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00001758
CAS号: 102-06-7
EINECS: 203-002-1
UN号: 3077
ADR: 9,III
Merck Index: 14,03324
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8adc7b94b0eea1015d1ceae98fd07e95d2ab42d7&vid=8adc7b94b0eea1015d1ceae98fd07e95d2ab42d7&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 152.0 deg-C
Solubility in Methanol almost transparency