1,2,3,4,5-五甲基环戊二烯 ≥93.0% (by GC)

供应商: TCI
Warning

Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

P1292-5ML P1292-25ML
TCIAP1292-5MLEA 0 CNY
TCIAP1292-5ML TCIAP1292-25ML
1,2,3,4,5-五甲基环戊二烯 ≥93.0% (by GC)
1,2,3,4,5-五甲基环戊二烯
分子式: C₁₀H₁₆
沸点: 60°C
密度: 0,87 g/cm³ (20 °C)
闪点: 44°C
存储条件: Ambient
MDL编号: MFCD00001354
CAS号: 4045-44-7
EINECS: 223-743-4
UN号: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dac43f66c73c5e376d452ac4b548111d43bee0c&vid=9dac43f66c73c5e376d452ac4b548111d43bee0c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR