5-甲基-1H-苯并三唑 ≥99.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 5-Methyl-1H-benzotriazole

M0249-25G M0249-500G
TCIAM0249-25GEA 0 CNY
TCIAM0249-25G TCIAM0249-500G
5-甲基-1H-苯并三唑 ≥99.0% (by titrimetric analysis)
5-甲基-1H-苯并三唑
分子式: C₇H₇N₃
分子量: 133.15 g/mol
沸点: 323°C
溶点: 85
存储条件: Refrigerator
MDL编号: MFCD00005702
CAS号: 136-85-6
EINECS: 205-265-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=884515db29fda1c7caf325738b97189256c0a020&vid=884515db29fda1c7caf325738b97189256c0a020&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 78.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR