1,3,3-三甲基-2-亚甲基吲哚啉 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-亚甲基-1,3,3-三甲基吲哚啉, Fischer 碱
T0879-500ML
T0879-25ML
TCIAT0879-500MLEA
0
CNY
TCIAT0879-500ML
TCIAT0879-25ML
1,3,3-三甲基-2-亚甲基吲哚啉 ≥96.0% (by GC)
1,3,3-三甲基-2-亚甲基吲哚啉
分子式:
C₁₂H₁₅N 沸点: 248°C 溶点: –10 °C 密度: 0,986 g/cm³ (20 °C) 闪点: 101°C 存储条件: Ambient |
MDL编号:
MFCD00005813 CAS号: 118-12-7 EINECS: 204-235-1 UN号: 3082 ADR: 9,III |
|
产品技术规格
Appearance | Red to Dark red to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...