二苯二硫醚 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
二硫化二苯
P0167-25G
P0167-500G
TCIAP0167-25GEA
0
CNY
TCIAP0167-25G
TCIAP0167-500G
二苯二硫醚 ≥99.0% (by GC)
二苯二硫醚
分子式:
C₁₂H₁₀S₂ 分子量: 218.34 g/mol 沸点: 310 |
MDL编号:
MFCD00003065 CAS号: 882-33-7 EINECS: 212-926-4 UN号: 3077 ADR: 9 III |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 59.0 to 62.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...