二丁基二硫 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
二硫二丁烷
D0224-25ML
D0224-500ML
TCIAD0224-25MLEA
0
CNY
TCIAD0224-25ML
TCIAD0224-500ML
二丁基二硫 ≥95.0% (by GC)
二丁基二硫
分子式:
C₈H₁₈S₂ 分子量: 178.36 g/mol 沸点: 88°C 溶点: -94 闪点: 93°C 存储条件: Ambient |
MDL编号:
MFCD00009467 CAS号: 629-45-8 EINECS: 211-091-3 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...