六氟磷酸钾 ≥95.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
P1023-25G
P1023-500G
TCIAP1023-25GEA
0
CNY
TCIAP1023-25G
TCIAP1023-500G
六氟磷酸钾 ≥95.0% (by titrimetric analysis)
六氟磷酸钾
分子式:
F₆KP 溶点: 575 °C 密度: 2,58 g/cm³ (20 °C) |
MDL编号:
MFCD00011412 CAS号: 17084-13-8 EINECS: 241-143-0 UN号: 3260 ADR: 8,II |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | 95.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...