白藜芦醇 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 5-[(1E)-2-(4-羟苯基)乙烯基]-1,3-苯二醇, 3,4′,5-三羟基-反式-二苯乙烯

R0071-1G R0071-5G
TCIAR0071-1GEA 0 CNY
TCIAR0071-1G TCIAR0071-5G
白藜芦醇 ≥99.0% (by GC)
白藜芦醇
分子式: C₁₄H₁₂O₃
分子量: 228.25 g/mol
溶点: 260
存储条件: Refrigerator
MDL编号: MFCD00133799
CAS号: 501-36-0
Merck Index: 14,08158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38b5115dabc40dd59d338f41e66aa9ddbc6cbab1&vid=38b5115dabc40dd59d338f41e66aa9ddbc6cbab1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR