三甲基氢醌 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2,3,5-三甲基-1,4-苯二酚, 2,3,5-三甲基对苯二酚

T0477-25G T0477-500G
TCIAT0477-25GEA 0 CNY
TCIAT0477-25G TCIAT0477-500G
三甲基氢醌 ≥98.0% (by GC, titration analysis)
三甲基氢醌
分子式: C₉H₁₂O₂
分子量: 152.19 g/mol
沸点: 295
存储条件: Ambient
MDL编号: MFCD00002346
CAS号: 700-13-0
EINECS: 211-838-3
UN号: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be38cfa985fc85393509aee8528aea7097bc29d5&vid=be38cfa985fc85393509aee8528aea7097bc29d5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Redox method) min. 98.0 %
Melting point 168.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR