胡椒醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
3,4-亚甲二氧基苯甲醇, 5-羟甲基苯并-1,3-二噁环戊烷, 3,4-亚甲二氧苄醇
P0457-25G
P0457-500G
TCIAP0457-25GEA
0
CNY
TCIAP0457-25G
TCIAP0457-500G
胡椒醇 ≥98.0% (by GC)
胡椒醇
分子式:
C₈H₈O₃ 沸点: 157°C 溶点: 51…54 °C |
MDL编号:
MFCD00005836 CAS号: 495-76-1 EINECS: 207-808-4 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...