二乙二醇 ≥99.5% (by GC)

供应商: TCI
Warning

Synonyms: 二(羟乙基)醚, 二甘醇, 2,2′-氧代二乙醇, 2-羟基乙基醚

D0495-100G D0495-500G
TCIAD0495-100GEA 0 CNY
TCIAD0495-100G TCIAD0495-500G
二乙二醇 ≥99.5% (by GC)
二乙二醇
分子式: C₄H₁₀O₃
分子量: 106.12 g/mol
沸点: 245°C
溶点: –7°C
闪点: 124°C (closed cup)
存储条件: Ambient
MDL编号: MFCD00002882
CAS号: 111-46-6
EINECS: 203-872-2
Merck Index: 14,03119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47fa5190d36dd34147a57881c675663f97d14cc&vid=a47fa5190d36dd34147a57881c675663f97d14cc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR