2-苯氧基乙醇 ≥98.5% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
乙二醇一苯醚, 苯基乙二醇
P0115-25G
P0115-500G
TCIAP0115-25GEA
0
CNY
TCIAP0115-25G
TCIAP0115-500G
2-苯氧基乙醇 ≥98.5% (by GC)
2-苯氧基乙醇
分子式:
C₆H₅OCH₂CH₂OH 分子量: 138.17 g/mol 沸点: 237°C 溶点: 13 闪点: 127°C (closed cup) 存储条件: Ambient |
MDL编号:
MFCD00002857 CAS号: 122-99-6 EINECS: 204-589-7 Merck Index: 14,07257 |
|
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...