4-甲氧基吲哚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Methoxy-1H-indole, Indol-4-yl methyl ether
M1250-1G
M1250-5G
TCIAM1250-1GEA
0
CNY
TCIAM1250-1G
TCIAM1250-5G
4-甲氧基吲哚 ≥98.0% (by GC)
4-甲氧基吲哚
分子式:
C₉H₉NO 分子量: 147.18 g/mol 沸点: 181°C |
MDL编号:
MFCD00009737 CAS号: 4837-90-5 |
|
产品技术规格
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 72.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...