2,4-二氨基嘧啶 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
Pyrimidine-2,4-diamine, Pyrimidine-2,4-diyltriamine
D2120-25G
D2120-5G
TCIAD2120-25GEA
0
CNY
TCIAD2120-25G
TCIAD2120-5G
2,4-二氨基嘧啶 ≥98.0% (by GC, titration analysis)
2,4-二氨基嘧啶
分子式:
C₄H₆N₄ |
MDL编号:
MFCD00038023 CAS号: 156-81-0 EINECS: 205-862-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...