磺胺酸 ≥99.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-氨基苯磺酸, 对氨基苯磺酸
S0120-25G
S0120-500G
TCIAS0120-25GEA
0
CNY
TCIAS0120-25G
TCIAS0120-500G
磺胺酸 ≥99.0% (by HPLC, titration analysis)
磺胺酸
分子式:
4-(H₂N)C₆H₄SO₃H 分子量: 173.19 g/mol 沸点: 363 溶点: 288°C 存储条件: Ambient |
MDL编号:
MFCD00007886 CAS号: 121-57-3 EINECS: 204-482-5 Merck Index: 14,08926 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in Na2CO3 | almost transparency |
Nitrous salt | max. 0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...