2-甲基-3-硝基苯胺 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
2-氨基-6-硝基甲苯, 3-硝基-邻甲苯胺
N0455-25G
N0455-250G
TCIAN0455-25GEA
0
CNY
TCIAN0455-25G
TCIAN0455-250G
2-甲基-3-硝基苯胺 ≥98.0% (by GC)
2-甲基-3-硝基苯胺
分子式:
C₇H₈N₂O₂ 分子量: 152.15 g/mol 沸点: 305°C 密度: 0.33 存储条件: Ambient |
MDL编号:
MFCD00007731 CAS号: 603-83-8 EINECS: 210-059-6 UN号: 2660 ADR: 6.1 III |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 91.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...