2-氨基-3-甲基吡啶 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-甲基-2-氨基吡啶

A0400-25G A0400-500G
TCIAA0400-25GEA 0 CNY
TCIAA0400-25G TCIAA0400-500G
2-氨基-3-甲基吡啶 ≥98.0% (by GC)
2-氨基-3-甲基吡啶
分子式: C₆H₈N₂
分子量: 108.14 g/mol
沸点: 221.1
存储条件: Ambient
MDL编号: MFCD00006320
CAS号: 1603-40-3
EINECS: 216-501-4
UN号: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02ac60fb48f013b0b66a2d9cd3fbb0e99bf5de31&vid=02ac60fb48f013b0b66a2d9cd3fbb0e99bf5de31&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 30.0 to 33.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR