2-氨基嘌呤 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-AP

A1111-1G A1111-200MG
TCIAA1111-1GEA 0 CNY
TCIAA1111-1G TCIAA1111-200MG
2-氨基嘌呤 ≥98.0% (by HPLC, titration analysis)
2-氨基嘌呤
分子式: C₅H₅N₅
分子量: 135.13 g/mol
溶点: 278°C
存储条件: Refrigerator
MDL编号: MFCD00005566
CAS号: 452-06-2
EINECS: 207-197-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dc7e1488a6cdad45d180bcc6cdaea4e9f8e499c&vid=2dc7e1488a6cdad45d180bcc6cdaea4e9f8e499c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR