异硫氰酸丁酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Butyl isothiocyanate

I0249-25ML I0249-250ML
TCIAI0249-25MLEA 0 CNY
TCIAI0249-25ML TCIAI0249-250ML
异硫氰酸丁酯 ≥98.0% (by GC)
异硫氰酸丁酯
分子式: C₅H₉NS
分子量: 115.2 g/mol
沸点: 66
闪点: 66°C
存储条件: Ambient
MDL编号: MFCD00004824
CAS号: 592-82-5
EINECS: 209-770-4
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee87ffefd7bf2a2efa734ad1839838845a1cb2d4&vid=ee87ffefd7bf2a2efa734ad1839838845a1cb2d4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR