肼二盐酸盐 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride

H0170-25G H0170-500G
TCIAH0170-25GEA 0 CNY
TCIAH0170-25G TCIAH0170-500G
肼二盐酸盐 ≥98.0% (by titrimetric analysis)
肼二盐酸盐
分子式: H₆Cl₂N₂
分子量: 104.97 g/mol
溶点: 189
MDL编号: MFCD00064543
CAS号: 5341-61-7
EINECS: 226-283-2
UN号: 3290
ADR: 6.1 II
Merck Index: 14,04770
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8baf560896824970acc24fcaa21202dfa78f940b&vid=8baf560896824970acc24fcaa21202dfa78f940b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR