3,4-二氟苯硫酚 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3,4-Difluorobenzene-1-thiol, 3,4-Difluorobenzenethiol, 1,2-Difluoro-4-mercaptobenzene, 3,4-Difluorothiophenol
D4121-25G
D4121-5G
TCIAD4121-25GEA
0
CNY
TCIAD4121-25G
TCIAD4121-5G
3,4-二氟苯硫酚 ≥97.0% (by GC)
3,4-二氟苯硫酚
分子式:
C₆H₄F₂S 分子量: 146.16 g/mol 沸点: 70°C |
MDL编号:
MFCD03094279 CAS号: 60811-24-7 UN号: 3336 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...