五氟丙酸酐 ≥95.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
全氟丙酸酐, PFPA
P0566-25G
P0566-5G
TCIAP0566-25GEA
0
CNY
TCIAP0566-25G
TCIAP0566-5G
五氟丙酸酐 ≥95.0% (by GC, titration analysis)
五氟丙酸酐
分子式:
C₆F₁₀O₃ 沸点: 80°C 密度: 1,571 g/cm³ (25 °C) 存储条件: Ambient |
MDL编号:
MFCD00000429 CAS号: 356-42-3 EINECS: 206-604-2 UN号: 1760 ADR: 8,III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Anilin Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...