2-氟-5-硝基苯甲酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

F0391-25G F0391-5G
TCIAF0391-25GEA 0 CNY
TCIAF0391-25G TCIAF0391-5G
2-氟-5-硝基苯甲酸 ≥98.0% (by GC, titration analysis)
2-氟-5-硝基苯甲酸
分子式: C₇H₄FNO₄
分子量: 185.11 g/mol
MDL编号: MFCD00134238
CAS号: 7304-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2db40f626c46ecb10b3ce54347d0f04275d1bbe3&vid=2db40f626c46ecb10b3ce54347d0f04275d1bbe3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR