5-氟-1-茚酮 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-Fluoro-2,3-dihydro-1H-inden-1-one
F0376-1G
F0376-5G
TCIAF0376-1GEA
0
CNY
TCIAF0376-1G
TCIAF0376-5G
5-氟-1-茚酮 ≥96.0% (by GC)
5-氟-1-茚酮
分子式:
C₉H₇FO 分子量: 150.15 g/mol 沸点: 114°C 存储条件: Refrigerator |
MDL编号:
MFCD00041031 CAS号: 700-84-5 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 37.0 to 40.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...