2-氟联苯 ≥97.0% (by GC)

供应商: TCI

Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1

F0265-25G F0265-5G
TCIAF0265-25GEA 0 CNY
TCIAF0265-25G TCIAF0265-5G
2-氟联苯 ≥97.0% (by GC)
2-氟联苯
分子式: C₁₂H₉F
分子量: 172.2 g/mol
沸点: 248°C
MDL编号: MFCD00000317
CAS号: 321-60-8
EINECS: 206-290-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bb3ebe69c9b7296d32577b4204882f991cfbba7&vid=7bb3ebe69c9b7296d32577b4204882f991cfbba7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 73.0 to 76.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR