2,2-Difluoro-1,3-benzodioxole ≥98.0% (通过GC)

供应商: TCI

D4051-25G D4051-5G
TCIAD4051-25GEA 0 CNY
TCIAD4051-25G TCIAD4051-5G
2,2-Difluoro-1,3-benzodioxole ≥98.0% (通过GC)

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR