硫代乙酰胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 乙硫酰胺

T0187-25G T0187-500G
TCIAT0187-25GEA 0 CNY
TCIAT0187-25G TCIAT0187-500G
硫代乙酰胺 ≥98.0% (by GC, titration analysis)
硫代乙酰胺
分子式: CH₃CSNH₂
溶点: 113…114 °C
密度: 1,336 g/cm³ (20 °C)
MDL编号: MFCD00008070
CAS号: 62-55-5
EINECS: 200-541-4
Merck Index: 14,09319
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dc87ed5e7383129d8274be2d7724e27cffe71f5&vid=8dc87ed5e7383129d8274be2d7724e27cffe71f5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 111.0 to 114.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR