2,3-吡啶二羧酸酐 ≥95.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
喹啉酸酐
P1222-25G
P1222-5G
TCIAP1222-25GEA
0
CNY
TCIAP1222-25G
TCIAP1222-5G
2,3-吡啶二羧酸酐 ≥95.0% (by titrimetric analysis)
2,3-吡啶二羧酸酐
分子式:
C₇H₃NO₃ 分子量: 149.11 g/mol 沸点: 360 溶点: 146 |
MDL编号:
MFCD00005915 CAS号: 699-98-9 EINECS: 211-834-1 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (Methanolysis Method) | min. 95.0 % |
Melting point | 135.0 to 140.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...