(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid
C1953-1G
C1953-5G
TCIAC1953-1GEA
0
CNY
TCIAC1953-1G
TCIAC1953-5G
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
分子式:
C₈H₁₂O₄ 分子量: 172.18 g/mol |
MDL编号:
MFCD00062973 CAS号: 46022-05-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 95.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 180.0 to 185.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...